missing translation for 'onlineSavingsMsg'
Learn More

2-Methylquinoline 98.0+%, TCI America™

Catalog No. Q000525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q000525G 25 g
Q0005100G 100 g
Q0005500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Q000525G Supplier TCI America Supplier No. Q000525G
Only null left

Chemical Identifiers

CAS 91-63-4
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006756
InChI Key SMUQFGGVLNAIOZ-UHFFFAOYSA-N
Synonym quinaldine, chinaldine, khinaldin, quinoline, 2-methyl, 2-methyl-quinoline, methylquinoline, 2-methylchinolin, ccris 1155, 2-methylchinoline, 2-methylchinolin czech
PubChem CID 7060
IUPAC Name 2-methylquinoline
SMILES CC1=CC=C2C=CC=CC2=N1

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 247°C
Quantity 25 g
Formula Weight 143.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methylquinoline
EINECSNumber (5)-0795&(5)-0796
RTECSNumber UZ9625000
TSCA Yes