missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylquinoline-6-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America M262525G
Specifications
| 2-Methylquinoline-6-carboxylic Acid | |
| 263°C | |
| C11H9NO2 | |
| 25 g | |
| IZONZQFTYGVOOO-UHFFFAOYSA-N | |
| 2-methylquinoline-6-carboxylic acid | |
| 604483 | |
| ≥98.0% (HPLC,T) |
| 635-80-3 | |
| Yellow | |
| MFCD00748504 | |
| 2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid | |
| CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O | |
| 187.198 | |
| 187.20 | |
| Crystalline Powder |
Chemical Identifiers
| 635-80-3 | |
| 187.198 | |
| IZONZQFTYGVOOO-UHFFFAOYSA-N | |
| 604483 | |
| CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O |
| C11H9NO2 | |
| MFCD00748504 | |
| 2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid | |
| 2-methylquinoline-6-carboxylic acid |
Safety and Handling
TSCA : No