missing translation for 'onlineSavingsMsg'
Learn More

2-Methylquinoline-6-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7133333
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
M26255G 5 g
1 options

Catalog No. M26255G

Supplier: TCI America M26255G

Only null left

Chemical Identifiers

CAS 635-80-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.198
MDL Number MFCD00748504
InChI Key IZONZQFTYGVOOO-UHFFFAOYSA-N
Synonym 2-methyl-6-quinolinecarboxylic acid, 6-quinolinecarboxylic acid, 2-methyl, 2-methyl-6-quinolinecarboxylicacid, 2-methylquinoline-6-carboxylicacid, acmc-1b4d8, quinaldine-6-carboxylic acid, 6-carboxy-2-methyl quinoline, ksc352o8f, 2-methyl-quinoline-6-carboxylic acid
PubChem CID 604483
IUPAC Name 2-methylquinoline-6-carboxylic acid
SMILES CC1=NC2=C(C=C1)C=C(C=C2)C(=O)O

Specifications

Melting Point 263°C
Color Yellow
Quantity 5 g
Formula Weight 187.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylquinoline-6-carboxylic Acid