Learn More
2-Methylpyridine-4-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 4021-11-8 |
|---|---|
| Molecular Formula | C7H7NO2 |
| Molecular Weight (g/mol) | 137.14 |
| InChI Key | PMDHIMMPXRSDML-UHFFFAOYSA-N |
| Synonym | 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl |
| PubChem CID | 2734672 |
| IUPAC Name | pyrazole-1-carboximidamide;hydrochloride |
| SMILES | CC1=NC=CC(=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC443490010
|
Thermo Scientific Chemicals
443490010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC443490050
|
Thermo Scientific Chemicals
443490050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 4021-11-8 | |
| 137.14 | |
| 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl | |
| pyrazole-1-carboximidamide;hydrochloride |
| C7H7NO2 | |
| PMDHIMMPXRSDML-UHFFFAOYSA-N | |
| 2734672 | |
| CC1=NC=CC(=C1)C(O)=O |
Specifications
| 4021-11-8 | |
| Glass Bottle | |
| 1g | |
| PMDHIMMPXRSDML-UHFFFAOYSA-N | |
| pyrazole-1-carboximidamide;hydrochloride | |
| 2734672 | |
| 97% |
| 96% min. (HPLC) | |
| C7H7NO2 | |
| 1h-pyrazole-1-carboxamidine hydrochloride, 1h-pyrazole-1-carboximidamide hydrochloride, 1-amidinopyrazole hydrochloride, praxadine, pyrazole-1-carboxamidine hydrochloride, pyrazole-1-carboximidamide hydrochloride, pyrazole-1-carboxamidine hcl, 1h-pyrazole-1-carboxamidinehydrochloride, praxadine hydrochloride, 1h-pyrazole-1-carboxamidine hcl | |
| CC1=NC=CC(=C1)C(O)=O | |
| 137.14 | |
| 137.14 | |
| 2-Methylpyridine-4-carboxylic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only