accessibility menu, dialog, popup

UserName

CAS RN 4021-11-8

CH3NOHO

CAS RN 4021-11-8

IUPAC Name: 2-methylpyridine-4-carboxylic acid
Synonyms: 2-methylpyridine-4-carboxylic acid
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: PMDHIMMPXRSDML-UHFFFAOYSA-N
SMILES: CC1=NC=CC(=C1)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)