missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpyridine-4-carboxylic acid, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Assay Percent Range | 96% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Synonym | 2-methylisonicotinic acid, 2-methyl-4-pyridinecarboxylic acid, 2-methyl-isonicotinic acid, 4-pyridinecarboxylic acid, 2-methyl, 4-carboxy-2-methylpyridine, methylisonicotinic acid 2-, 2-methylisonicotinicacid, acmc-20a0jw, 2-methylisonicotinic aicd, 6-methylpyridine-4-carboxylic acid |
| SMILES | CC1=NC=CC(=C1)C(=O)O |
| IUPAC Name | 2-methylpyridine-4-carboxylic acid |
| PubChem CID | 2762821 |
| Formula Weight | 137.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Methylpyridine-4-carboxylic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only