missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpyridine-3-boronic acid hydrochloride, 97%

Catalog No. AC430390010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430390010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430390010 Supplier Thermo Scientific Chemicals Supplier No. 430390010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1072952-34-1 | C6H9BClNO2 | 173.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072952-34-1
Molecular Formula C6H9BClNO2
Molecular Weight (g/mol) 173.40
MDL Number MFCD06659489
InChI Key OAMCCRLFDPJOPN-UHFFFAOYSA-N
Synonym 2-methylpyridin-3-yl boronic acid hydrochloride, 2-picoline-3-boronic acid hcl, 2-methylpyridin-3-ylboronic acid hydrochloride, 2-picoline-3-boronic acid hydrochloride, 2-methylpyridine-3-boronic acid hydrochloride, 2-picoline-3-boronic acid hydrochloride salt, 2-methylpyridin-3-yl-3-boronic acid hydrochloride, 2-methylpyridin-3-yl boronic acid-hydrogen chloride 1/1
PubChem CID 44119808
IUPAC Name (2-methylpyridin-3-yl)boronic acid;hydrochloride
SMILES Cl.CC1=C(C=CC=N1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 181.0°C to 186.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 173.41
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2-Methylpyridine-3-boronic acid hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only