CAS RN 1072952-34-1
IUPAC Name:
(2-methylpyridin-3-yl)boronic acid hydrochloride
Synonyms:
2-methylpyridin-3-ylboronic acid hydrochloride
Molecular Weight (g/mol):
173.4
Molecular Formula:
C6H9BClNO2
InChi Key:
OAMCCRLFDPJOPN-UHFFFAOYSA-N
SMILES:
Cl.CC1=C(C=CC=N1)B(O)O
1
–
1
of
1
results
1
Promotions Available
2-Methylpyridine-3-boronic acid hydrochloride, 97%
2-Methylpyridine-3-boronic acid hydrochloride, 97%
CAS: 1072952-34-1 Molecular Formula: C6H9BClNO2 Molecular Weight (g/mol): 173.40 MDL Number: MFCD06659489 InChI Key: OAMCCRLFDPJOPN-UHFFFAOYSA-N Synonym: 2-methylpyridin-3-yl boronic acid hydrochloride,2-picoline-3-boronic acid hcl,2-methylpyridin-3-ylboronic acid hydrochloride,2-picoline-3-boronic acid hydrochloride,2-methylpyridine-3-boronic acid hydrochloride,2-picoline-3-boronic acid hydrochloride salt,2-methylpyridin-3-yl-3-boronic acid hydrochloride,2-methylpyridin-3-yl boronic acid-hydrogen chloride 1/1 PubChem CID: 44119808 IUPAC Name: (2-methylpyridin-3-yl)boronic acid;hydrochloride SMILES: Cl.CC1=C(C=CC=N1)B(O)O