missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpiperidine 98.0+%, TCI America™
Supplier: TCI America P0443100ML
Specifications
| 2-Methylpiperidine | |
| Yellow | |
| C6H13N | |
| 100 mL | |
| 2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline | |
| CC1CCCCN1 | |
| 99.18 | |
| 99.18 | |
| Liquid |
| 109-05-7 | |
| 118°C | |
| MFCD00005982 | |
| 1993 | |
| NNWUEBIEOFQMSS-UHFFFAOYNA-N | |
| 2-methylpiperidine | |
| 7974 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 109-05-7 | |
| 99.18 | |
| NNWUEBIEOFQMSS-UHFFFAOYNA-N | |
| 7974 | |
| CC1CCCCN1 |
| C6H13N | |
| MFCD00005982 | |
| 2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline | |
| 2-methylpiperidine |
Safety and Handling
EINECSNumber : (5)-0768
RTECSNumber : TN1240000
TSCA : Yes