accessibility menu, dialog, popup

UserName

CH3HN

CAS RN 109-05-7

IUPAC Name: 2-methylpiperidine
Synonyms: 2-methylpiperidine
Molecular Weight (g/mol): 99.18
Molecular Formula: C6H13N
InChi Key: NNWUEBIEOFQMSS-UHFFFAOYNA-N
SMILES: CC1CCCCN1
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)