missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpiperidine 98.0+%, TCI America™

Catalog No. P044325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P044325ML 25 mL
P0443100ML 100 mL
P0443500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P044325ML Supplier TCI America Supplier No. P044325ML

Chemical Identifiers

CAS 109-05-7
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00005982
InChI Key NNWUEBIEOFQMSS-UHFFFAOYNA-N
Synonym 2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline
PubChem CID 7974
IUPAC Name 2-methylpiperidine
SMILES CC1CCCCN1

Specifications

Color Yellow
Boiling Point 118°C
Quantity 25 mL
UN Number 1993
Formula Weight 99.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylpiperidine
EINECSNumber (5)-0768
RTECSNumber TN1240000
TSCA Yes