missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpiperidine, 98+%

Catalog No. AAL02567AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL02567AP 500 mL
AAL02567AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL02567AP Supplier Thermo Scientific Chemicals Supplier No. L02567AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 109-05-7 | C6H13N | 99.18 g/mol

2-Methylpiperidine, is used for the oxidation of secondary amines to nitrones by means of H2O2 catalyzed by Sodium tungsten oxide dihydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylpiperidine, is used for the oxidation of secondary amines to nitrones by means of H2O2 catalyzed by Sodium tungsten oxide dihydrate.

Solubility
It is easily soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 109-05-7
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00005982
InChI Key NNWUEBIEOFQMSS-UHFFFAOYNA-N
Synonym 2-pipecoline, pipicoline, alpha-pipecoline, piperidine, 2-methyl, alpha-pipecolin, pipecoline, alpha, 2-methyl-piperidine, alpha-methylpiperidine, .alpha.-pipecolin, .alpha.-pipecoline
PubChem CID 7974
IUPAC Name 2-methylpiperidine
SMILES CC1CCCCN1

Specifications

Melting Point -4°C
Density 0.842
Boiling Point 118°C to 119°C
Flash Point 8°C (46°F)
Odor Amine-like
Refractive Index 1.447
Quantity 500 mL
UN Number UN2733
Beilstein 79804
Solubility Information It is easily soluble in water.
Formula Weight 99.18
Percent Purity ≥98%
Chemical Name or Material 2-Methylpiperidine
Show More Show Less
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 203-642-1
RTECSNumber TN1240000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only