missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpiperazine 99.0+%, TCI America™

Catalog No. M1179500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1179500G 500 g
M117925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1179500G Supplier TCI America Supplier No. M1179500G

Chemical Identifiers

CAS 109-07-9
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00005954
InChI Key JOMNTHCQHJPVAZ-UHFFFAOYSA-N
Synonym piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
PubChem CID 66057
IUPAC Name 2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 153°C
Quantity 500 g
UN Number 2925
Formula Weight 100.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylpiperazine
EINECSNumber (5)-3829
RTECSNumber TM1225020
TSCA Yes