accessibility menu, dialog, popup

UserName

CAS RN 109-07-9

IUPAC Name: 2-methylpiperazine
Synonyms: piperazine, 2-methyl-
Molecular Weight (g/mol): 100.17
Molecular Formula: C5H12N2
InChi Key: JOMNTHCQHJPVAZ-UHFFFAOYNA-N
SMILES: CC1CNCCN1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)