missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylpentane, ≥99.5% (GC), MilliporeSigma™ Supelco™
Description
Featured Industries: Petroleum
Specifications
Specifications
| CAS | 107-83-5 |
| Ignition Point | 583°F |
| Vapor Pressure | 6.77 psi (37.7°C) |
| Vapor Density | 3 (vs. air) |
| Density | 0.653 g/mL (at 25°C) |
| Flash Point | -7°C - closed cup |
| Physical Form | Neat |
| Linear Formula | CH3 CH2 CH2 CH(CH3)2 |
| MDL Number | MFCD00009406 |
| Melting Point | -154°C |
| Show More |