missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylnaphtho[1,2-d]thiazole 98.0+%, TCI America™
$158.44 - $158.44
Chemical Identifiers
| CAS | 2682-45-3 |
|---|---|
| Molecular Formula | C12H9NS |
| Molecular Weight (g/mol) | 199.27 |
| MDL Number | MFCD00004951 |
| InChI Key | OUXMJRMYZCEVKO-UHFFFAOYSA-N |
| Synonym | 2-methylnaphtho 1,2-d thiazole, 2-methyl-beta-naphthothiazole, 2-methylnaphtho 1,2-d 1,3 thiazole, naphtho 1,2-d thiazole, 2-methyl, 2-methyl-b-naphthylthiazole, naphth 1,2-d thiazole, 2-methyl, 2-methyl-.beta.-naphthothiazole, 2-methyinaphtho 1,2-d thiazole, 2-methylbenzo e 1,3 benzothiazole |
| PubChem CID | 75892 |
| IUPAC Name | 2-methylnaphtho[1,2-d][1,3]thiazole |
| SMILES | CC1=NC2=C(S1)C=CC1=CC=CC=C21 |
Chemical Identifiers
| 2682-45-3 | |
| 199.27 | |
| OUXMJRMYZCEVKO-UHFFFAOYSA-N | |
| 75892 | |
| CC1=NC2=C(S1)C=CC1=CC=CC=C21 |
| C12H9NS | |
| MFCD00004951 | |
| 2-methylnaphtho 1,2-d thiazole, 2-methyl-beta-naphthothiazole, 2-methylnaphtho 1,2-d 1,3 thiazole, naphtho 1,2-d thiazole, 2-methyl, 2-methyl-b-naphthylthiazole, naphth 1,2-d thiazole, 2-methyl, 2-methyl-.beta.-naphthothiazole, 2-methyinaphtho 1,2-d thiazole, 2-methylbenzo e 1,3 benzothiazole | |
| 2-methylnaphtho[1,2-d][1,3]thiazole |
Specifications
| 2682-45-3 | |
| Yellow | |
| C12H9NS | |
| 5 g | |
| OUXMJRMYZCEVKO-UHFFFAOYSA-N | |
| 2-methylnaphtho[1,2-d][1,3]thiazole | |
| 75892 | |
| ≥98.0% (GC) | |
| 2-Methylnaphtho[1,2-d]thiazole |
| 96°C | |
| 168°C | |
| MFCD00004951 | |
| 2-methylnaphtho 1,2-d thiazole, 2-methyl-beta-naphthothiazole, 2-methylnaphtho 1,2-d 1,3 thiazole, naphtho 1,2-d thiazole, 2-methyl, 2-methyl-b-naphthylthiazole, naphth 1,2-d thiazole, 2-methyl, 2-methyl-.beta.-naphthothiazole, 2-methyinaphtho 1,2-d thiazole, 2-methylbenzo e 1,3 benzothiazole | |
| CC1=NC2=C(S1)C=CC1=CC=CC=C21 | |
| 199.27 | |
| 199.27 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3439
TSCA : Yes