accessibility menu, dialog, popup

UserName

CAS RN 2682-45-3

IUPAC Name: 2-methylnaphtho[1,2-d][1,3]thiazole
Synonyms: 2-methylnaphtho[1,2-d][1,3]thiazole
Molecular Weight (g/mol): 199.27
Molecular Formula: C12H9NS
InChi Key: OUXMJRMYZCEVKO-UHFFFAOYSA-N
SMILES: CC1=NC2=C(S1)C=CC1=CC=CC=C21
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results