missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnaphtho[1,2-d]thiazole 98.0+%, TCI America™

Catalog No. M12875G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12875G Supplier TCI America Supplier No. M12875G

Chemical Identifiers

CAS 2682-45-3
Molecular Formula C12H9NS
Molecular Weight (g/mol) 199.27
MDL Number MFCD00004951
InChI Key OUXMJRMYZCEVKO-UHFFFAOYSA-N
Synonym 2-methylnaphtho 1,2-d thiazole, 2-methyl-beta-naphthothiazole, 2-methylnaphtho 1,2-d 1,3 thiazole, naphtho 1,2-d thiazole, 2-methyl, 2-methyl-b-naphthylthiazole, naphth 1,2-d thiazole, 2-methyl, 2-methyl-.beta.-naphthothiazole, 2-methyinaphtho 1,2-d thiazole, 2-methylbenzo e 1,3 benzothiazole
PubChem CID 75892
IUPAC Name 2-methylnaphtho[1,2-d][1,3]thiazole
SMILES CC1=NC2=C(S1)C=CC1=CC=CC=C21

Specifications

Melting Point 96°C
Color Yellow
Boiling Point 168°C
Quantity 5 g
Formula Weight 199.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylnaphtho[1,2-d]thiazole
EINECSNumber (5)-3439
TSCA Yes