missing translation for 'onlineSavingsMsg'
Learn More

2-Methylnaphthalene 96.0+%, TCI America™

Catalog No. M037225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M037225G 25 g
M0372500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M037225G Supplier TCI America Supplier No. M037225G

Chemical Identifiers

CAS 91-57-6
Molecular Formula C11H10
Molecular Weight (g/mol) 142.201
MDL Number MFCD00004118
InChI Key QIMMUPPBPVKWKM-UHFFFAOYSA-N
Synonym naphthalene, 2-methyl, beta-methylnaphthalene, .beta.-methylnaphthalene, naphthalene, beta-methyl, unii-s8mcx3c16h, 2-methyl naphthalene, 2-methyl-naphthalene, beta-methyl naphthalenes, s8mcx3c16h, naphtalene,2-methyl mfc11 h10
PubChem CID 7055
ChEBI CHEBI:50720
IUPAC Name 2-methylnaphthalene
SMILES CC1=CC2=CC=CC=C2C=C1

Specifications

Color White-Yellow
Boiling Point 241°C
Quantity 25 g
UN Number 3077
Formula Weight 142.20
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methylnaphthalene
EINECSNumber (4)-0080
RTECSNumber QJ9635000
TSCA Yes