Learn More
2-Methylnaphthalene, 96%
CAS: 91-57-6 | C11H10 | 142.201 g/mol
Supplier: Thermo Scientific Chemicals L0245922
Description
The main uses of methylnaphthalene are as a raw material for dyestuff dispersants and heat transfer oils, and as a solvent for agricultural chemical. It has been used as an indicator of smoke exposure in food and packaging materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe main uses of methylnaphthalene are as a raw material for dyestuff dispersants and heat transfer oils, and as a solvent for agricultural chemical. It has been used as an indicator of smoke exposure in food and packaging materials.
Solubility
Insoluble in water. Soluble in alcohol, chloroform, methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 2-Methylnaphthalene | |
| 31°C to 35°C | |
| 241°C to 243°C | |
| Characteristic | |
| MFCD00004118 | |
| UN3077 | |
| naphthalene, 2-methyl, beta-methylnaphthalene, .beta.-methylnaphthalene, naphthalene, beta-methyl, unii-s8mcx3c16h, 2-methyl naphthalene, 2-methyl-naphthalene, beta-methyl naphthalenes, s8mcx3c16h, naphtalene,2-methyl mfc11 h10 | |
| QIMMUPPBPVKWKM-UHFFFAOYSA-N | |
| 2-methylnaphthalene | |
| 7055 | |
| 142.2 |
| 91-57-6 | |
| 1.001 | |
| 97°C (206°F) | |
| C11H10 | |
| 100 g | |
| 906859 | |
| Insoluble in water. Soluble in alcohol,chloroform,methanol. | |
| CC1=CC2=CC=CC=C2C=C1 | |
| 142.201 | |
| CHEBI:50720 | |
| 97% |
Chemical Identifiers
| 91-57-6 | |
| 142.201 | |
| QIMMUPPBPVKWKM-UHFFFAOYSA-N | |
| 7055 | |
| 2-methylnaphthalene |
| C11H10 | |
| MFCD00004118 | |
| naphthalene, 2-methyl, beta-methylnaphthalene, .beta.-methylnaphthalene, naphthalene, beta-methyl, unii-s8mcx3c16h, 2-methyl naphthalene, 2-methyl-naphthalene, beta-methyl naphthalenes, s8mcx3c16h, naphtalene,2-methyl mfc11 h10 | |
| CHEBI:50720 | |
| CC1=CC2=CC=CC=C2C=C1 |
Safety and Handling
GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 202-078-3
RTECSNumber : QJ9635000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only