Learn More
2-Methylnaphthalene, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 127170025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methylnaphthalene | |
| 91-57-6 | |
| 32°C to 36°C | |
| 1.0100g/mL | |
| 97°C | |
| 95% min. (GC) | |
| C11H10 | |
| MFCD00004118 | |
| 05, 567 | |
| naphthalene, 2-methyl, beta-methylnaphthalene, .beta.-methylnaphthalene, naphthalene, beta-methyl, unii-s8mcx3c16h, 2-methyl naphthalene, 2-methyl-naphthalene, beta-methyl naphthalenes, s8mcx3c16h, naphtalene,2-methyl mfc11 h10 | |
| QIMMUPPBPVKWKM-UHFFFAOYSA-N | |
| 2-methylnaphthalene | |
| 7055 | |
| 142.2 | |
| Crystalline Low Melting Solid |
| 96% | |
| 96 | |
| White | |
| 241°C to 242°C | |
| Authentic | |
| Glass bottle | |
| C10H7CH3 | |
| 2.5 kg | |
| 1.01 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
| CC1=CC2=CC=CC=C2C=C1 | |
| 142.2 | |
| CHEBI:50720 | |
| 96% |
Chemical Identifiers
| 91-57-6 | |
| 142.2 | |
| QIMMUPPBPVKWKM-UHFFFAOYSA-N | |
| 7055 | |
| 2-methylnaphthalene |
| C11H10 | |
| MFCD00004118 | |
| naphthalene, 2-methyl, beta-methylnaphthalene, .beta.-methylnaphthalene, naphthalene, beta-methyl, unii-s8mcx3c16h, 2-methyl naphthalene, 2-methyl-naphthalene, beta-methyl naphthalenes, s8mcx3c16h, naphtalene,2-methyl mfc11 h10 | |
| CHEBI:50720 | |
| CC1=CC2=CC=CC=C2C=C1 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-078-3
RTECSNumber : QJ9635000
TSCA : TSCA
RUO â Research Use Only