Learn More
2-Methylindoline, 99+%
CAS: 6872-06-6 | C9H11N | 133.19 g/mol
$57.20 - $255.39
Chemical Identifiers
| CAS | 6-6-6872 |
|---|---|
| Molecular Formula | C9H11N |
| Molecular Weight (g/mol) | 133.19 |
| MDL Number | MFCD00005706 |
| InChI Key | QRWRJDVVXAXGBT-UHFFFAOYNA-N |
| Synonym | 2-methylindoline, 2-methyl-2,3-dihydro-1h-indole, 2,3-dihydro-2-methylindole, 1h-indole, 2,3-dihydro-2-methyl, indoline, 2-methyl, 2-methyl-2,3-dihydroindole, dihydroindole, 2-methyl, alpha-methyldihydroindole, .alpha.-methyldihydroindole, 2,3-dihydro-2-methyl-1h-indole |
| PubChem CID | 23305 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC275880050
|
Thermo Scientific Chemicals
275880050 |
5 mL | Glass bottle |
Each for $57.20
|
|
||||
|
AC275881000
|
Thermo Scientific Chemicals
275881000 |
100 mL | Glass bottle |
Each for $255.39
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6-6-6872 | |
| 133.19 | |
| QRWRJDVVXAXGBT-UHFFFAOYNA-N | |
| 23305 |
| C9H11N | |
| MFCD00005706 | |
| 2-methylindoline, 2-methyl-2,3-dihydro-1h-indole, 2,3-dihydro-2-methylindole, 1h-indole, 2,3-dihydro-2-methyl, indoline, 2-methyl, 2-methyl-2,3-dihydroindole, dihydroindole, 2-methyl, alpha-methyldihydroindole, .alpha.-methyldihydroindole, 2,3-dihydro-2-methyl-1h-indole |
Specifications
| 6-6-6872 | |
| 100.0 | |
| Yellow to Brown | |
| 227°C (1013.0 Mbar) | |
| Authentic | |
| Glass bottle | |
| 1.5680 to 1.57 | |
| 5 mL | |
| 1.02 | |
| Solubility in water: negligible | |
| 133.19 | |
| 133.19 | |
| Liquid |
| 99.0 | |
| -51°C | |
| 1.0200g/mL | |
| 103°C | |
| 99+% | |
| C9H11N | |
| MFCD00005706 | |
| 20, 279 | |
| 2-methylindoline, 2-methyl-2,3-dihydro-1h-indole, 2,3-dihydro-2-methylindole, 1h-indole, 2,3-dihydro-2-methyl, indoline, 2-methyl, 2-methyl-2,3-dihydroindole, dihydroindole, 2-methyl, alpha-methyldihydroindole, .alpha.-methyldihydroindole, 2,3-dihydro-2-methyl-1h-indole | |
| QRWRJDVVXAXGBT-UHFFFAOYNA-N | |
| 23305 | |
| 99+% | |
| 2-Methylindoline, 99+% |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 229-971-0
RTECSNumber : NM1926350
TSCA : TSCA
RUO – Research Use Only