Learn More
2-Methylindoline, 99+%
CAS: 6872-06-6 | C9H11N | 133.19 g/mol
Supplier: Thermo Scientific Chemicals 275881000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methylindoline | |
| 6-6-6872 | |
| 100.0 | |
| Yellow to Brown | |
| 227°C (1013.0 Mbar) | |
| Authentic | |
| Glass bottle | |
| 1.5680 to 1.57 | |
| 100 mL | |
| 1.02 | |
| Solubility in water: negligible | |
| 133.19 | |
| 133.19 | |
| Liquid |
| 99+% | |
| 99.0 | |
| -51°C | |
| 1.0200g/mL | |
| 103°C | |
| 99+% | |
| C9H11N | |
| MFCD00005706 | |
| 20, 279 | |
| 2-methylindoline, 2-methyl-2,3-dihydro-1h-indole, 2,3-dihydro-2-methylindole, 1h-indole, 2,3-dihydro-2-methyl, indoline, 2-methyl, 2-methyl-2,3-dihydroindole, dihydroindole, 2-methyl, alpha-methyldihydroindole, .alpha.-methyldihydroindole, 2,3-dihydro-2-methyl-1h-indole | |
| QRWRJDVVXAXGBT-UHFFFAOYNA-N | |
| 23305 | |
| 99+% |
Chemical Identifiers
| 6-6-6872 | |
| 133.19 | |
| QRWRJDVVXAXGBT-UHFFFAOYNA-N | |
| 23305 |
| C9H11N | |
| MFCD00005706 | |
| 2-methylindoline, 2-methyl-2,3-dihydro-1h-indole, 2,3-dihydro-2-methylindole, 1h-indole, 2,3-dihydro-2-methyl, indoline, 2-methyl, 2-methyl-2,3-dihydroindole, dihydroindole, 2-methyl, alpha-methyldihydroindole, .alpha.-methyldihydroindole, 2,3-dihydro-2-methyl-1h-indole |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 229-971-0
RTECSNumber : NM1926350
TSCA : TSCA
RUO – Research Use Only