missing translation for 'onlineSavingsMsg'
Learn More

2-Methylindole 99.0+%, TCI America™

Catalog No. M0346500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0346500G 500 g
M034625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0346500G Supplier TCI America Supplier No. M0346500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-20-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005616
InChI Key BHNHHSOHWZKFOX-UHFFFAOYSA-N
Synonym 2-methylindole, 1h-indole, 2-methyl, indole, 2-methyl, 2-methyl indole, unii-i7cn58827i, 2-methylindol, methylketole, methyl indole, 2-methyl-indole, pubchem7504
PubChem CID 7224
ChEBI CHEBI:49402
IUPAC Name 2-methyl-1H-indole
SMILES CC1=CC2=CC=CC=C2N1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 272°C
Quantity 500 g
Formula Weight 131.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylindole
EINECSNumber (5)-0131
RTECSNumber NM0345000
TSCA Yes