Learn More
2-Methylindole, 98+%
CAS: 95-20-5 | C9H9N | 131.178 g/mol
Supplier: Thermo Scientific Chemicals A107640B
Description
2-Methylindole is used as a reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction, Friedel-Crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, Michael addition reactions and in synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylindole is used as a reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction, Friedel-Crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, Michael addition reactions and in synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
Specifications
| 2-Methylindole | |
| 56°C to 60°C | |
| 272°C to 273°C | |
| C9H9N | |
| 1000 g | |
| Air and light sensitive | |
| Insoluble in water. | |
| CC1=CC2=CC=CC=C2N1 | |
| 131.178 | |
| CHEBI:49402 | |
| ≥98% |
| 95-20-5 | |
| 1.07 | |
| 141°C (285°F) | |
| MFCD00005616 | |
| 109781 | |
| 2-methylindole, 1h-indole, 2-methyl, indole, 2-methyl, 2-methyl indole, unii-i7cn58827i, 2-methylindol, methylketole, methyl indole, 2-methyl-indole, pubchem7504 | |
| BHNHHSOHWZKFOX-UHFFFAOYSA-N | |
| 2-methyl-1H-indole | |
| 7224 | |
| 131.18 |
Chemical Identifiers
| 95-20-5 | |
| 131.178 | |
| BHNHHSOHWZKFOX-UHFFFAOYSA-N | |
| 7224 | |
| 2-methyl-1H-indole |
| C9H9N | |
| MFCD00005616 | |
| 2-methylindole, 1h-indole, 2-methyl, indole, 2-methyl, 2-methyl indole, unii-i7cn58827i, 2-methylindol, methylketole, methyl indole, 2-methyl-indole, pubchem7504 | |
| CHEBI:49402 | |
| CC1=CC2=CC=CC=C2N1 |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 202-398-3
RTECSNumber : NM0345000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only