missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylimidazole 98.0+%, TCI America™
Supplier: TCI America M0345100G
Specifications
| 2-Methylimidazole | |
| 144°C | |
| 268°C | |
| MFCD00005190 | |
| 3263 | |
| LXBGSDVWAMZHDD-UHFFFAOYSA-N | |
| 2-methyl-1H-imidazole | |
| 12749 | |
| ≥97.0% (GC,T) |
| 693-98-1 | |
| White-Yellow | |
| C4H6N2 | |
| 100 g | |
| 2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107 | |
| CC1=NC=CN1 | |
| 82.11 | |
| 82.11 | |
| Crystalline Powder |
Chemical Identifiers
| 693-98-1 | |
| 82.11 | |
| LXBGSDVWAMZHDD-UHFFFAOYSA-N | |
| 12749 | |
| CC1=NC=CN1 |
| C4H6N2 | |
| MFCD00005190 | |
| 2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107 | |
| 2-methyl-1H-imidazole |
Safety and Handling
EINECSNumber : (5)-0382
RTECSNumber : NI7175000
TSCA : Yes