missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazole 98.0+%, TCI America™

Catalog No. p-7132113
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M034525G 25 g
M0345100G 100 g
M0345500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M034525G Supplier TCI America Supplier No. M034525G
Only null left

Chemical Identifiers

CAS 693-98-1
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005190
InChI Key LXBGSDVWAMZHDD-UHFFFAOYSA-N
Synonym 2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
PubChem CID 12749
IUPAC Name 2-methyl-1H-imidazole
SMILES CC1=NC=CN1

Specifications

Melting Point 144°C
Color White-Yellow
Boiling Point 268°C
Quantity 25 g
UN Number 3263
Formula Weight 82.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylimidazole
EINECSNumber (5)-0382
RTECSNumber NI7175000
TSCA Yes