missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazo[4,5-c]pyridine, 96%

Catalog No. AAL19131MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19131MD 250 mg
AAL1913103 1 g
AAL1913106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL19131MD Supplier Thermo Scientific Chemicals Supplier No. L19131MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63604-59-1 | C7H7N3 | 133.154 g/mol

2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylimidazo[4,5-c]pyridine is used as a antagonist against platelet-activating factor (PAF).

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 63604-59-1
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.154
MDL Number MFCD03426025
InChI Key JWQVOMJORQXNME-UHFFFAOYSA-N
Synonym 2-methyl-3h-imidazo 4,5-c pyridine, 2-methyl-1h-imidazo 4,5-c pyridine, 2-methylimidazo 4,5-c pyridine, 8-methyl-3-deazapurine, 8-methyl-3,7,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene, acmc-20anq0, 2-methyl-5-azabenzimidazole, 3h-imidazo 4,5-c pyridine, 2-methyl, 1h-imidazo 4,5-c pyridine, 2-methyl
PubChem CID 373077
IUPAC Name 2-methyl-3H-imidazo[4,5-c]pyridine
SMILES CC1=NC2=C(N1)C=NC=C2

Specifications

Melting Point 176°C to 178°C
Quantity 250 mg
Beilstein 3234
Solubility Information Insoluble in water.
Formula Weight 133.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Methylimidazo[4,5-c]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only