CAS RN 63604-59-1
IUPAC Name:
2-methyl-3H-imidazo[4,5-c]pyridine
Synonyms:
2-methyl-3H-imidazo[4,5-c]pyridine
Molecular Weight (g/mol):
133.15
Molecular Formula:
C7H7N3
InChi Key:
JWQVOMJORQXNME-UHFFFAOYSA-N
SMILES:
CC1=NC2=CC=NC=C2N1
1
–
1
of
1
results
1
Promotions Available
2-Methylimidazo[4,5-c]pyridine, 96%
2-Methylimidazo[4,5-c]pyridine, 96%
CAS: 63604-59-1 Molecular Formula: C7H7N3 Molecular Weight (g/mol): 133.154 MDL Number: MFCD03426025 InChI Key: JWQVOMJORQXNME-UHFFFAOYSA-N Synonym: 2-methyl-3h-imidazo 4,5-c pyridine,2-methyl-1h-imidazo 4,5-c pyridine,2-methylimidazo 4,5-c pyridine,8-methyl-3-deazapurine,8-methyl-3,7,9-triazabicyclo 4.3.0 nona-2,4,7,10-tetraene,acmc-20anq0,2-methyl-5-azabenzimidazole,3h-imidazo 4,5-c pyridine, 2-methyl,1h-imidazo 4,5-c pyridine, 2-methyl PubChem CID: 373077 IUPAC Name: 2-methyl-3H-imidazo[4,5-c]pyridine SMILES: CC1=NC2=C(N1)C=NC=C2