missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylhexanoic Acid 98.0+%, TCI America™
Supplier: TCI America M094725ML
| Quantity | 25 mL |
|---|
Chemical Identifiers
| C7H13O2 | |
| MFCD00002674 | |
| 2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl | |
| (2R)-2-methylhexanoate |
Specifications
| 2-Methylhexanoic Acid | |
| Yellow | |
| C7H13O2 | |
| MFCD00002674 | |
| 2-methylcaproic acid, hexanoic acid, 2-methyl, methylhexanoic acid, 2-methyl hexanoic acid, 2-hexanecarboxylic acid, alpha-methylcaproic acid, 2-methyl-hexanoic acid, fema no. 3191, .alpha.-methylcaproic acid, hexanoic acid, methyl | |
| CCCC[C@@H](C)C([O-])=O | |
| 129.18 | |
| 130.19 | |
| Liquid |
| 4536-23-6 | |
| 72°C | |
| 25 mL | |
| 3265 | |
| CVKMFSAVYPAZTQ-ZCFIWIBFSA-M | |
| (2R)-2-methylhexanoate | |
| 20653 | |
| ≥98.0% (GC,T) |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : Yes