missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylhexane 99.0+%, TCI America™
Supplier: TCI America M023125ML
Specifications
| 2-Methylhexane | |
| Colorless | |
| C7H16 | |
| 25 mL | |
| hexane, 2-methyl, isoheptane, unii-yi4679p2sp, pentane, dimethyl, isoheptanes, 2-methyl hexane, 2-methylhexane, acmc-1b07e, 5-methylhexan-2-yl, 5-methylhexan-3-yl | |
| CCCCC(C)C | |
| 100.21 | |
| 100.21 | |
| Liquid |
| 591-76-4 | |
| 90°C | |
| MFCD00009487 | |
| 3295 | |
| GXDHCNNESPLIKD-UHFFFAOYSA-N | |
| 2-methylhexane | |
| 11582 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 591-76-4 | |
| 100.21 | |
| GXDHCNNESPLIKD-UHFFFAOYSA-N | |
| 11582 | |
| CCCCC(C)C |
| C7H16 | |
| MFCD00009487 | |
| hexane, 2-methyl, isoheptane, unii-yi4679p2sp, pentane, dimethyl, isoheptanes, 2-methyl hexane, 2-methylhexane, acmc-1b07e, 5-methylhexan-2-yl, 5-methylhexan-3-yl | |
| 2-methylhexane |
Safety and Handling
EINECSNumber : (2)-0007
RTECSNumber : MO3871500
TSCA : Yes