missing translation for 'onlineSavingsMsg'
Learn More

2-Methylhexane 99.0+%, TCI America™

Catalog No. p-7132073
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02315ML 5 mL
M023125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M02315ML Supplier TCI America Supplier No. M02315ML
Only null left

Chemical Identifiers

CAS 591-76-4
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009487
InChI Key GXDHCNNESPLIKD-UHFFFAOYSA-N
Synonym hexane, 2-methyl, isoheptane, unii-yi4679p2sp, pentane, dimethyl, isoheptanes, 2-methyl hexane, 2-methylhexane, acmc-1b07e, 5-methylhexan-2-yl, 5-methylhexan-3-yl
PubChem CID 11582
IUPAC Name 2-methylhexane
SMILES CCCCC(C)C

Specifications

Color Colorless
Boiling Point 90°C
Quantity 5 mL
UN Number 3295
Formula Weight 100.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylhexane
EINECSNumber (2)-0007
RTECSNumber MO3871500
TSCA Yes