missing translation for 'onlineSavingsMsg'
Learn More

2-Methylglutaric Acid 98.0+%, TCI America™

Catalog No. p-7132650
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13005G 5 g
M130025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13005G Supplier TCI America Supplier No. M13005G
Only null left

Chemical Identifiers

CAS 617-62-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00002661
InChI Key AQYCMVICBNBXNA-UHFFFAOYSA-N
Synonym 2-methylglutaric acid, alpha-methylglutaric acid, pentanedioic acid, 2-methyl, glutaric acid, 2-methyl, 2-methyl-glutaric acid, 2-methylglutarate, .alpha.-methylglutaric acid, 2-methyl glutaric acid, 2-methyl-pentanedioic acid, 2-methylpentanedioate
PubChem CID 12046
ChEBI CHEBI:68567
IUPAC Name 2-methylpentanedioic acid
SMILES CC(CCC(=O)O)C(=O)O

Specifications

Melting Point 77°C
Boiling Point 205°C
Quantity 5 g
Formula Weight 146.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylglutaric Acid