accessibility menu, dialog, popup

UserName

CH3HOOOHO

CAS RN 617-62-9

IUPAC Name: 2-methylpentanedioic acid
Synonyms: α-methylglutaric acid (5-L-glutamyl)-L-glutamate ion(2-) α-methylene glutarate α-methylene glutarate, ion(2-)
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: AQYCMVICBNBXNA-UHFFFAOYNA-N
SMILES: CC(CCC(O)=O)C(O)=O
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)