missing translation for 'onlineSavingsMsg'
Learn More

2-Methylglutaric acid, 98%

Catalog No. p-7081728
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1182306 5 g
AAA1182314 25 g
AAA1182322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1182306 Supplier Thermo Scientific Chemicals Supplier No. A1182306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 617-62-9 | C6H10O4 | 146.142 g/mol

2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Incompatible with bases and oxidizing agents.

Chemical Identifiers

CAS 617-62-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00002661
InChI Key AQYCMVICBNBXNA-UHFFFAOYSA-N
Synonym 2-methylglutaric acid, alpha-methylglutaric acid, pentanedioic acid, 2-methyl, glutaric acid, 2-methyl, 2-methyl-glutaric acid, 2-methylglutarate, .alpha.-methylglutaric acid, 2-methyl glutaric acid, 2-methyl-pentanedioic acid, 2-methylpentanedioate
PubChem CID 12046
ChEBI CHEBI:68567
IUPAC Name 2-methylpentanedioic acid
SMILES CC(CCC(=O)O)C(=O)O

Specifications

Melting Point 74°C to 78°C
Boiling Point 214°C to 215°C (22 mmHg)
Quantity 5 g
Beilstein 1723219
Solubility Information Soluble in water.
Formula Weight 146.14
Percent Purity 98%
Chemical Name or Material 2-Methylglutaric acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
EINECSNumber 210-521-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only