missing translation for 'onlineSavingsMsg'
Learn More

2-Methylfuran, 98+%, stab.

CAS: 534-22-5 | C5H6O | 82.102 g/mol

$132.56 - $409.55

Chemical Identifiers

CAS 534-22-5
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.102
MDL Number MFCD00003248
InChI Key VQKFNUFAXTZWDK-UHFFFAOYSA-N
Synonym methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran
PubChem CID 10797
IUPAC Name 2-methylfuran
SMILES CC1=CC=CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB23692AK
View Documents
Thermo Scientific Chemicals
B23692AK
250 mL
Each for $132.56
Only null left
 
AAB23692AU
View Documents
Thermo Scientific Chemicals
B23692AU
1000 mL
Each for $409.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

534-22-5
82.102
VQKFNUFAXTZWDK-UHFFFAOYSA-N
10797
CC1=CC=CO1
C5H6O
MFCD00003248
methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran
2-methylfuran

Specifications

534-22-5
0.910
-30°C (-22°F)
C5H6O
MFCD00003248
UN2301
methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran
CC1=CC=CO1
82.102
82.1
2-Methylfuran, stab. with 150-400ppm BHT
°C to 89°C
63°C to 65°C
98+%
1.433
250 mL
103733
VQKFNUFAXTZWDK-UHFFFAOYSA-N
2-methylfuran
10797
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H300-H330-H341-H319

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c

H225-H301+H311+H331

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-METHYLFURAN

EINECSNumber : 208-594-5

RTECSNumber : LU2625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only