Learn More
2-(Methyldithio)isobutyraldehyde, 95%
CAS: 67952-60-7 | C5H10OS2 | 150.25 g/mol
$372.70 - $372.70
Chemical Identifiers
| CAS | 67952-60-7 |
|---|---|
| Molecular Formula | C5H10OS2 |
| Molecular Weight (g/mol) | 150.25 |
| InChI Key | VLBWEJJOETYCSE-UHFFFAOYSA-N |
| Synonym | 2-methyldithio isobutyraldehyde, 2-methyl-2-methyldithio propanal, unii-3d5g4az6l5, propanal, 2-methyl-2-methyldithio, 2-methyldithio-isobutyraldehyde, alpha-methyldithio isobutyraldehyde, 2-methyl-2-methyldisulfanyl propanal, 2-methyl-2-methyldithio propionaldehyde, 2-methyldisulfanyl isobutyraldehyde, cocoa propanal |
| PubChem CID | 106217 |
| IUPAC Name | 2-methyl-2-(methyldisulfanyl)propanal |
| SMILES | CC(C)(C=O)SSC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC265150010
|
Thermo Scientific Chemicals
265150010 |
1 g | Glass bottle |
Each for $372.70
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 67952-60-7 | |
| 150.25 | |
| 2-methyldithio isobutyraldehyde, 2-methyl-2-methyldithio propanal, unii-3d5g4az6l5, propanal, 2-methyl-2-methyldithio, 2-methyldithio-isobutyraldehyde, alpha-methyldithio isobutyraldehyde, 2-methyl-2-methyldisulfanyl propanal, 2-methyl-2-methyldithio propionaldehyde, 2-methyldisulfanyl isobutyraldehyde, cocoa propanal | |
| 2-methyl-2-(methyldisulfanyl)propanal |
| C5H10OS2 | |
| VLBWEJJOETYCSE-UHFFFAOYSA-N | |
| 106217 | |
| CC(C)(C=O)SSC |
Specifications
| 67952-60-7 | |
| 1.1000g/mL | |
| 72°C | |
| 94% min. (GC) | |
| C5H10OS2 | |
| 1 g | |
| 2-methyldithio isobutyraldehyde, 2-methyl-2-methyldithio propanal, unii-3d5g4az6l5, propanal, 2-methyl-2-methyldithio, 2-methyldithio-isobutyraldehyde, alpha-methyldithio isobutyraldehyde, 2-methyl-2-methyldisulfanyl propanal, 2-methyl-2-methyldithio propionaldehyde, 2-methyldisulfanyl isobutyraldehyde, cocoa propanal | |
| VLBWEJJOETYCSE-UHFFFAOYSA-N | |
| 2-methyl-2-(methyldisulfanyl)propanal | |
| 106217 | |
| 95% | |
| 2-(Methyldithio)isobutyraldehyde |
| Yellow | |
| 46.0°C (1.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.5190 to 1.5230 | |
| 1.1 | |
| Solubility in water: practically insoluble | |
| CC(C)(C=O)SSC | |
| 150.25 | |
| 150.25 | |
| Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
EINECSNumber : 267-914-1
RUO – Research Use Only