missing translation for 'onlineSavingsMsg'
Learn More

2-Methyldibenzo[f,h]quinoxaline 98.0+%, TCI America™

Catalog No. M3005200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3005200MG 200 mg
M30051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M3005200MG Supplier TCI America Supplier No. M3005200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 536753-86-3
Molecular Formula C17H12N2
Molecular Weight (g/mol) 244.297
MDL Number MFCD16038624
InChI Key DCPGBPKLXYETTA-UHFFFAOYSA-N
PubChem CID 20720413
IUPAC Name 3-methylphenanthro[9,10-b]pyrazine
SMILES CC1=CN=C2C3=CC=CC=C3C4=CC=CC=C4C2=N1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 200 mg
Formula Weight 244.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyldibenzo[f,h]quinoxaline