accessibility menu, dialog, popup

UserName

H3CNN

CAS RN 536753-86-3

IUPAC Name: 2-methyl-1,4-diazatriphenylene
Synonyms: 2-methyl-1,4-diazatriphenylene
Molecular Weight (g/mol): 244.3
Molecular Formula: C17H12N2
InChi Key: DCPGBPKLXYETTA-UHFFFAOYSA-N
SMILES: CC1=CN=C2C3=CC=CC=C3C3=CC=CC=C3C2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results