missing translation for 'onlineSavingsMsg'
Learn More

2-Methylcyclopentane-1,3-dione, 98%

Catalog No. AAA1393606
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1393606 5 g
AAA1393614 25 g
AAA1393622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1393606 Supplier Thermo Scientific Chemicals Supplier No. A1393606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 765-69-5 | C6H8O2 | 112.128 g/mol

2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylcyclopentane-1,3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone. The alkylation of 2-methylcyclopentane-1,3-dione (A) with 2-(1-naphthyl)ethyl bromide gives only the O-alkylation product. Mixtures of C- and O-alkyl derivatives are obtained. Michael addition of 2-methylcyclopentane-1,3-dione to methyl vinyl ketone in water gives 2-methyl-2-(3-oxo- butyl)-1,3-cyclopentanedione (3), an important intermediate of natural product chemistry. 2-Methylcyclopentane-1, 3-dione is an efficient synthon for the synthesis of (±)-α-cuparenone.

Solubility
Solubility in hot Methanol (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 765-69-5
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001406
InChI Key HXZILEQYFQYQCE-UHFFFAOYSA-N
Synonym 2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
PubChem CID 13005
IUPAC Name 2-methylcyclopentane-1,3-dione
SMILES CC1C(=O)CCC1=O

Specifications

Melting Point 212°C to 216°C
Quantity 5 g
Beilstein 1237255
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material 2-Methylcyclopentane-1,3-dione
Hazard Category H500
EINECSNumber 212-153-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only