accessibility menu, dialog, popup

UserName

CAS RN 765-69-5

IUPAC Name: 2-methylcyclopentane-1,3-dione
Synonyms: 2-methylcyclopentane-1,3-dione
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: HXZILEQYFQYQCE-UHFFFAOYSA-N
SMILES: CC1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)