missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutyraldehyde, ≥95% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019892
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019892 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019892 Supplier Merck Emd Millipore Supplier No. 61904100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 96-17-3
Density 0.806 g/mL (at 20°C)
Flash Point 4°C
Physical Form Neat
Linear Formula C2H5 CH(CH3)CHO
MDL Number MFCD00006984
Boiling Point 90°C to 92°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 86.13
Percent Purity ≥95% (GC)
Grade Analytical Standard
Show More Show Less