missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbutane, Reagent Grade, ≥99%, Honeywell™
Reagent Grade,≥99%
Supplier: Honeywell Inc M326314X1L
Description
- ≥95% for general laboratory use.
Specifications
| 2-Methylbutane | |
| -160°C | |
| 0.62g/mL at 25°C | |
| −51°C (−59.8°F) | |
| Characteristic | |
| C5H12 | |
| MFCD00009338 | |
| UN1265 | |
| isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
| QWTDNUCVQCZILF-UHFFFAOYSA-N | |
| 2-methylbutane | |
| 2.6 (vs air) | |
| CHEBI:30362 | |
| 11.17psi (20°C) |
| 78-78-4 | |
| Colorless | |
| 28°C (1013 hPa) | |
| 788°F | |
| Glass bottle | |
| CH3CH2CH(CH3)2 | |
| 4 x 1 L | |
| 1730723 | |
| Insoluble in water | |
| CCC(C)C | |
| 72.15 | |
| 6556 | |
| 72.15g/mol | |
| ≥99% |
Chemical Identifiers
| 78-78-4 | |
| 72.15 | |
| QWTDNUCVQCZILF-UHFFFAOYSA-N | |
| 6556 | |
| 2-methylbutane |
| C5H12 | |
| MFCD00009338 | |
| isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
| CHEBI:30362 | |
| CCC(C)C |
Safety and Handling
P210-P261-P273-P301 + P310-P331
DOTInformation : class 3 - PG 1 - Pentanes
RTECSNumber : 201-142-8