missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, Reagent Grade, ≥99%, Honeywell™

Catalog No. p-7120071
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 x 1 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60048074 500 mL Glass bottle
60048070 1 L Glass bottle
60048071 2.5 L Glass bottle
60048073 4 x 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60048074 Supplier Honeywell Chemicals Supplier No. M32631500ML
Only null left

Reagent Grade,≥99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Melting Point -160°C
Color Colorless
Density 0.62g/mL at 25°C
Boiling Point 28°C (1013 hPa)
Flash Point −51°C (−59.8°F)
Ignition Point 788°F
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)2
Quantity 500 mL
UN Number UN1265
Beilstein 1730723
Solubility Information Insoluble in water
Vapor Density 2.6 (vs air)
Vapor Pressure 11.17psi (20°C)
Formula Weight 72.15g/mol
Percent Purity ≥99%
Chemical Name or Material 2-Methylbutane
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331
DOTInformation class 3 - PG 1 - Pentanes
RTECSNumber 201-142-8