missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzylamine 98.0+%, TCI America™

Catalog No. p-7132455
Change view
Click to view available options
Quantity:
5 mL
25 mL
Catalog No. Quantity
M09425ML 5 mL
M094225ML 25 mL
2 options

Catalog No. M09425ML

Supplier: TCI America M09425ML

Only null left

Chemical Identifiers

CAS 89-93-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00008112
InChI Key CJAAPVQEZPAQNI-UHFFFAOYSA-N
Synonym 2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
PubChem CID 6993
IUPAC Name (2-methylphenyl)methanamine
SMILES CC1=CC=CC=C1CN

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 mL
UN Number 2735
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methylbenzylamine
RTECSNumber DP5800000
TSCA Yes