missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzylamine, 96%, Thermo Scientific™

Chemical Identifiers

CAS 89-93-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008112
InChI Key CJAAPVQEZPAQNI-UHFFFAOYSA-N
Synonym 2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
PubChem CID 6993
IUPAC Name (2-methylphenyl)methanamine
SMILES CC1=CC=CC=C1CN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC155630050
View Documents Promotion Details
N/A 5g Glass bottle N/A
Only null left
 
AC155630250
View Documents Promotion Details
N/A 25g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

89-93-0
121.18
CJAAPVQEZPAQNI-UHFFFAOYSA-N
6993
CC1=CC=CC=C1CN
C8H11N
MFCD00008112
2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
(2-methylphenyl)methanamine

Specifications

89-93-0
100.0
196°C to 197°C
Authentic
C8H11N
CH3C6H4CH2NH2
12, 1106
0.97
CJAAPVQEZPAQNI-UHFFFAOYSA-N
(2-methylphenyl)methanamine
6993
96%
95.0
0.9700g/mL
83°C
96%
1.5415 to 1.5435
MFCD00008112
13,185
2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
CC1=CC=CC=C1CN
121.18
121.18
2-Methylbenzylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 201-952-1

SDS
Documents

Documents

RUO – Research Use Only