missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzylamine, 96%

Catalog No. AC155630250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155630250 25 g Glass bottle
AC155630050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC155630250 Supplier Thermo Scientific Chemicals Supplier No. 155630250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 89-93-0 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-93-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008112
InChI Key CJAAPVQEZPAQNI-UHFFFAOYSA-N
Synonym 2-methylbenzylamine, o-tolylmethanamine, o-methylbenzylamine, benzenemethanamine, 2-methyl, o-xylylamine, benzylamine, o-methyl, 2-methylphenyl methanamine, alpha-amino-o-xylene, alpha-amino-2-xylene, 1-2-methylphenyl methanamine
PubChem CID 6993
IUPAC Name (2-methylphenyl)methanamine
SMILES CC1=CC=CC=C1CN

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9700g/mL
Boiling Point 196°C to 197°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Refractive Index 1.5415 to 1.5435
Linear Formula CH3C6H4CH2NH2
Quantity 25 g
Beilstein 12, 1106
Fieser 13,185
Specific Gravity 0.97
Formula Weight 121.18
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Methylbenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 201-952-1

RUO – Research Use Only