missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzyl Alcohol 97.0+%, TCI America™

Catalog No. M088725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M088725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M088725G Supplier TCI America Supplier No. M088725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-95-2
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004622
InChI Key XPNGNIFUDRPBFJ-UHFFFAOYSA-N
Synonym 2-methylbenzyl alcohol, o-tolylmethanol, o-tolyl alcohol, 2-methylphenyl methanol, o-methylbenzyl alcohol, benzenemethanol, 2-methyl, toluene-methanol, benzyl alcohol, o-methyl, unii-7l3m6y04nc, alpha-hydroxy-o-xylene
PubChem CID 6994
ChEBI CHEBI:27724
IUPAC Name (2-methylphenyl)methanol
SMILES CC1=CC=CC=C1CO

Specifications

Melting Point 36°C
Color White
Boiling Point 219°C
Quantity 25 g
Formula Weight 122.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylbenzyl Alcohol