missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzyl alcohol, 98%

CAS: 89-95-2 | C8H10O | 122.167 g/mol

Supplier:  Thermo Scientific Chemicals A1269618

 View more versions of this product

Catalog No. AAA1269618


Only null left
Explore available promotions

Description

Description

2-Methylbenzyl alcohol is widely used strong mobile phase additive in HPLC. it can be used to produce 2-methyl-benzaldehyde at temperature of 20°C. It will need reagent pyridinium chlorochromate with reaction time of 10 min.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzyl alcohol is widely used strong mobile phase additive in HPLC. it can be used to produce 2-methyl-benzaldehyde at temperature of 20°C. It will need reagent pyridinium chlorochromate with reaction time of 10 min.

Solubility
Soluble in methanol 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents.
Specifications

Specifications

2-Methylbenzyl alcohol
33°C to 37°C
104°C (219°F)
1.5408
50 g
2-methylbenzyl alcohol, o-tolylmethanol, o-tolyl alcohol, 2-methylphenyl methanol, o-methylbenzyl alcohol, benzenemethanol, 2-methyl, toluene-methanol, benzyl alcohol, o-methyl, unii-7l3m6y04nc, alpha-hydroxy-o-xylene
XPNGNIFUDRPBFJ-UHFFFAOYSA-N
(2-methylphenyl)methanol
6994
122.17
89-95-2
219°C
C8H10O
MFCD00004622
1929783
Soluble in methanol 0.1g/mL.
CC1=CC=CC=C1CO
122.167
CHEBI:27724
98%

Chemical Identifiers

89-95-2
122.167
XPNGNIFUDRPBFJ-UHFFFAOYSA-N
6994
(2-methylphenyl)methanol
C8H10O
MFCD00004622
2-methylbenzyl alcohol, o-tolylmethanol, o-tolyl alcohol, 2-methylphenyl methanol, o-methylbenzyl alcohol, benzenemethanol, 2-methyl, toluene-methanol, benzyl alcohol, o-methyl, unii-7l3m6y04nc, alpha-hydroxy-o-xylene
CHEBI:27724
CC1=CC=CC=C1CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 201-954-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only