missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzyl alcohol, 98%

Catalog No. AAA1269609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1269609 10 g
AAA1269618 50 g
AAA1269630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1269609 Supplier Thermo Scientific Chemicals Supplier No. A1269609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 89-95-2 | C8H10O | 122.167 g/mol

2-Methylbenzyl alcohol is widely used strong mobile phase additive in HPLC. it can be used to produce 2-methyl-benzaldehyde at temperature of 20°C. It will need reagent pyridinium chlorochromate with reaction time of 10 min.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzyl alcohol is widely used strong mobile phase additive in HPLC. it can be used to produce 2-methyl-benzaldehyde at temperature of 20°C. It will need reagent pyridinium chlorochromate with reaction time of 10 min.

Solubility
Soluble in methanol 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from acids, acid chlorides, acid anhydrides, oxidizing agents.

Chemical Identifiers

CAS 89-95-2
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004622
InChI Key XPNGNIFUDRPBFJ-UHFFFAOYSA-N
Synonym 2-methylbenzyl alcohol, o-tolylmethanol, o-tolyl alcohol, 2-methylphenyl methanol, o-methylbenzyl alcohol, benzenemethanol, 2-methyl, toluene-methanol, benzyl alcohol, o-methyl, unii-7l3m6y04nc, alpha-hydroxy-o-xylene
PubChem CID 6994
ChEBI CHEBI:27724
IUPAC Name (2-methylphenyl)methanol
SMILES CC1=CC=CC=C1CO

Specifications

Melting Point 33°C to 37°C
Boiling Point 219°C
Flash Point 104°C (219°F)
Refractive Index 1.5408
Quantity 10 g
Beilstein 1929783
Solubility Information Soluble in methanol 0.1g/mL.
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2-Methylbenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-954-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only